Learn More
4-Bromobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals
Description
4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 272°C to 274°C |
Quantity | 10 g |
Beilstein | 3693458 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 222.52 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromobenzylamine hydrochloride |
Safety and Handling
Hazard Category | H302+H312-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 247-503-3 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.