Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobiphenyl, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
92-66-0
C12H9Br
233.11
MFCD00000100
PKJBWOWQJHHAHG-UHFFFAOYSA-N
4-bromobiphenyl,p-bromobiphenyl,4-bromo-1,1'-biphenyl,4-bromodiphenyl,1,1'-biphenyl, 4-bromo,4-biphenyl bromide,p-bromodiphenyl,biphenyl, 4-bromo,4-bromo-biphenyl,p-phenylbromobenzene
7101
1-bromo-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)Br
92-66-0
C12H9Br
233.11
MFCD00000100
PKJBWOWQJHHAHG-UHFFFAOYSA-N
4-bromobiphenyl,p-bromobiphenyl,4-bromo-1,1'-biphenyl,4-bromodiphenyl,1,1'-biphenyl, 4-bromo,4-biphenyl bromide,p-bromodiphenyl,biphenyl, 4-bromo,4-bromo-biphenyl,p-phenylbromobenzene
7101
1-bromo-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)Br
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 143°C
Color White to Yellow
Quantity 25g
Boiling Point 310.0°C
Melting Point 89.0°C to 92.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5C6H4Br
Beilstein 05, 580
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, ether and benzene
Formula Weight 233.11
Physical Form Crystalline Flakes
Percent Purity 99%
Chemical Name or Material 4-Bromobiphenyl, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 202-176-6

RTECSNumber : DV1750100

TSCA : TSCA