Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromobutyl acetate, 98+%

Click to view available options
Quantity:
10g
50g
250g
4753-59-7
C6H11BrO2
195.056
MFCD00000263
UOABIRUEGSGTSA-UHFFFAOYSA-N
4-bromobutylacetate,4-bromo-n-butyl acetate,1-butanol, 4-bromo-, acetate,acetic acid 4-bromobutyl ester,1-butanol, 4-bromo-, 1-acetate,4-bromo-1-acetoxybutane,1-butanol, 4-bromo-,1-acetate,bromobutyl acetate,4-acetoxy-butylbromide,4-acetoxybutyl bromide
78491
4-bromobutyl acetate
CC(=O)OCCCCBr
4753-59-7
C6H11BrO2
195.056
MFCD00000263
UOABIRUEGSGTSA-UHFFFAOYSA-N
4-bromobutylacetate,4-bromo-n-butyl acetate,1-butanol, 4-bromo-, acetate,acetic acid 4-bromobutyl ester,1-butanol, 4-bromo-, 1-acetate,4-bromo-1-acetoxybutane,1-butanol, 4-bromo-,1-acetate,bromobutyl acetate,4-acetoxy-butylbromide,4-acetoxybutyl bromide
78491
4-bromobutyl acetate
CC(=O)OCCCCBr
Density 1.348
Refractive Index 1.46
Flash Point 109°C (228°F)
Boiling Point 96°C to 97°C (15mmHg)
Quantity 50g
Beilstein 1749695
Formula Weight 195.07
Percent Purity ≥98%
Chemical Name or Material 4-Bromobutyl acetate

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 225-277-7

TSCA : Yes

Recommended Storage : Ambient temperatures