Alfa Aesar™ 4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%

Manufacturer: Alfa Aesar™ H27097MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27097MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 165881-36-7
    Molecular Formula C14H24BBrO2
    Formula Weight 315.06
    MDL Number MFCD09037500
    Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol 4-bromobutylboronate
    Chemical Name or Material 4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester

    Specifications

    Formula Weight 315.06
    Quantity 250mg
    Optical Rotation +18° (c=1 in Chloroform)
    Molecular Formula C14H24BBrO2
    MDL Number MFCD09037500
    CAS 165881-36-7
    Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol 4-bromobutylboronate
    Chemical Name or Material 4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit