Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromobutyronitrile, 97%

Manufacturer:  Alfa Aesar™ A1067430

Quantity 250g

 View more versions of this product

Catalog No. AAA1067430


  Check Availability
Add to cart

Specifications

Description

Applications
4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.

Solubility
Miscible with toluene and benzene. Immiscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Description & Specifications

Chemical Identifiers

5332-06-9
148.01
UN3276
1-Bromo-3-cyanopropane
Br(CH2)3CN
MFCD00001971
956577
4-Bromobutyronitrile

Specifications

5332-06-9
250g
Odorless
148.01
UN3276
1-Bromo-3-cyanopropane
97%
204° to 206°C
103°C (217°F)
Br(CH2)3CN
MFCD00001971
956577
4-Bromobutyronitrile

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : ET8925000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit