Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromoimidazole, 98%

Click to view available options
Quantity:
5g
25g
2302-25-2
C3H3BrN2
146.975
MFCD00047021
FHZALEJIENDROK-UHFFFAOYSA-N
4-bromo-1h-imidazole,4-bromoimidazole,1h-imidazole, 4-bromo,4 or 5-bromoimidazole,imidazole, 4-bromo,4-bromo-3h-imidazole,imidazole, 4 or 5-bromo-6ci,7ci,5-bromoimidazole,3kqm,4-bromo-imidazole
96125
5-bromo-1H-imidazole
C1=C(NC=N1)Br
2302-25-2
C3H3BrN2
146.975
MFCD00047021
FHZALEJIENDROK-UHFFFAOYSA-N
4-bromo-1h-imidazole,4-bromoimidazole,1h-imidazole, 4-bromo,4 or 5-bromoimidazole,imidazole, 4-bromo,4-bromo-3h-imidazole,imidazole, 4 or 5-bromo-6ci,7ci,5-bromoimidazole,3kqm,4-bromo-imidazole
96125
5-bromo-1H-imidazole
C1=C(NC=N1)Br
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Melting Point 131°C to 135°C
UN Number UN2811
Beilstein 1615
Formula Weight 146.97
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromoimidazole

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : NI3517450