Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoisophthalic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
6939-93-1
C8H5BrO4
245.028
MFCD00002404
MSQIEZXCNYUWHN-UHFFFAOYSA-N
4-bromoisophthalic acid,1,3-benzenedicarboxylic acid, 4-bromo,4-bromoisophthalicacid,4-bromo-1,3-benzenedicarboxylic acid,2-bromobenzene-1,5-dicarboxylic acid,pubchem12479,chemwish ic03530,ksc354c4d,msqiezxcnyuwhn-uhfffaoysa
81361
4-bromobenzene-1,3-dicarboxylic acid
C1=CC(=C(C=C1C(=O)O)C(=O)O)Br
6939-93-1
C8H5BrO4
245.028
MFCD00002404
MSQIEZXCNYUWHN-UHFFFAOYSA-N
4-bromoisophthalic acid,1,3-benzenedicarboxylic acid, 4-bromo,4-bromoisophthalicacid,4-bromo-1,3-benzenedicarboxylic acid,2-bromobenzene-1,5-dicarboxylic acid,pubchem12479,chemwish ic03530,ksc354c4d,msqiezxcnyuwhn-uhfffaoysa
81361
4-bromobenzene-1,3-dicarboxylic acid
C1=CC(=C(C=C1C(=O)O)C(=O)O)Br
Packaging Glass Bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 297.0°C to 299.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 09, 838
Formula Weight 245.03
Physical Form Crystalline Powder and Chunks
Percent Purity 97%
Chemical Name or Material 4-Bromoisophthalic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.