Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromoisoquinoline, 98%

Click to view available options
Quantity:
5g
25g
100g
1532-97-4
C9H6BrN
208.058
MFCD00006904
SCRBSGZBTHKAHU-UHFFFAOYSA-N
isoquinoline, 4-bromo,4-bromo-isoquinoline,4-isoquinolinyl bromide,4-bromisochinolin,4bromoisoquinoline,4-bromoisoquinoline,4-bromo isoquinoline,4-bromoisoquinoline?,pubchem6212,acmc-1bu1h
73743
4-bromoisoquinoline
C1=CC=C2C(=C1)C=NC=C2Br
1532-97-4
C9H6BrN
208.058
MFCD00006904
SCRBSGZBTHKAHU-UHFFFAOYSA-N
isoquinoline, 4-bromo,4-bromo-isoquinoline,4-isoquinolinyl bromide,4-bromisochinolin,4bromoisoquinoline,4-bromoisoquinoline,4-bromo isoquinoline,4-bromoisoquinoline?,pubchem6212,acmc-1bu1h
73743
4-bromoisoquinoline
C1=CC=C2C(=C1)C=NC=C2Br
Boiling Point 285°C
Melting Point 39°C to 43°C
Quantity 100g
Flash Point >110°C (230°F)
Beilstein 114431
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 4-Bromoisoquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 216-244-8

TSCA : No

Recommended Storage : Ambient temperatures