Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromomethyl-2(1H)-quinolinone

Click to view available options
Quantity:
1g
5g
4876-10-2
C10H8BrNO
238.08
MFCD03426152
BBAHJCUCNVVEQU-UHFFFAOYSA-N
4-bromomethyl-1,2-dihydroquinoline-2-one,4-bromomethyl quinolin-2 1h-one,4-bromomethyl-2 1h-quinolinone,4-bromomethyl-1,2-dihydroquinolin-2-one,4-bromomethyl-1h-quinolin-2-one,2 1h-quinolinone, 4-bromomethyl,4-bromomethyl-2-quinolinone,2-hydroxy-4-bromo methyl quinoline,4-bromomethyl hydroquinolin-2-one,4-bromomethylquinolinon
268296
4-(bromomethyl)-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)CBr
4876-10-2
C10H8BrNO
238.08
MFCD03426152
BBAHJCUCNVVEQU-UHFFFAOYSA-N
4-bromomethyl-1,2-dihydroquinoline-2-one,4-bromomethyl quinolin-2 1h-one,4-bromomethyl-2 1h-quinolinone,4-bromomethyl-1,2-dihydroquinolin-2-one,4-bromomethyl-1h-quinolin-2-one,2 1h-quinolinone, 4-bromomethyl,4-bromomethyl-2-quinolinone,2-hydroxy-4-bromo methyl quinoline,4-bromomethyl hydroquinolin-2-one,4-bromomethylquinolinon
268296
4-(bromomethyl)-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)CBr
Melting Point ~254°C (decomposition)
Quantity 1g
UN Number UN3261
Formula Weight 238.08
Chemical Name or Material 4-Bromomethyl-2(1H)-quinolinone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.