Alfa Aesar™ 4-Bromomethyl-6,7-dimethoxycoumarin, 95%

Fluorescent labeling reagent; Used in HPLC for the detection of carboxylic acids

Manufacturer: Alfa Aesar™ 4340202

Quantity 0.5g

 View more versions of this product

Catalog No. AA4340202

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88404-25-5
    Molecular Formula C12H11BrO4
    Formula Weight 299.12
    MDL Number MFCD00011570
    UN Number UN3261
    Beilstein 4750487
    Chemical Name or Material 4-Bromomethyl-6,7-dimethoxycoumarin

    Specifications

    Formula Weight 299.12
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 88404-25-5
    Physical Form Powder
    Molecular Formula C12H11BrO4
    UN Number UN3261
    Melting Point 200° to 203°C
    Beilstein 4750487
    Health Hazard 1 Danger
    MDL Number MFCD00011570
    Quantity 0.5g
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4-Bromomethyl-6,7-dimethoxycoumarin
    Assay 95%

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit