Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromomethyl)-7-methoxycoumarin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
35231-44-8
C11H9BrO3
269.08
MFCD00006869
CTENSLORRMFPDH-UHFFFAOYSA-N
4-bromomethyl-7-methoxycoumarin,4-bromomethyl-7-methoxy-2h-chromen-2-one,br-mmc,ccris 7996,4-bromomethyl-7-methoxychromen-2-one,2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy,4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran,acmc-1agnd,4-brommethyl-7-methoxy-2h-chromen-2-on,4-bromomethyl-7-methoxy coumarin
121894
4-(bromomethyl)-7-methoxychromen-2-one
COC1=CC2=C(C=C1)C(=CC(=O)O2)CBr
35231-44-8
C11H9BrO3
269.08
MFCD00006869
CTENSLORRMFPDH-UHFFFAOYSA-N
4-bromomethyl-7-methoxycoumarin,4-bromomethyl-7-methoxy-2h-chromen-2-one,br-mmc,ccris 7996,4-bromomethyl-7-methoxychromen-2-one,2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy,4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran,acmc-1agnd,4-brommethyl-7-methoxy-2h-chromen-2-on,4-bromomethyl-7-methoxy coumarin
121894
4-(bromomethyl)-7-methoxychromen-2-one
COC1=CC2=C(C=C1)C(=CC(=O)O2)CBr
Melting Point 212.0°C to 217.0°C
Color Beige or White to Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble in water. Other solubilities: solubel in most common organic solvents
Formula Weight 269.08
Physical Form Powder and/or Chunks
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)-7-methoxycoumarin
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-448-3