Alfa Aesar™ 4-(Bromomethyl)benzenesulfonyl chloride, 95%

Manufacturer: Alfa Aesar™ A1012003

Quantity 1g

 View more versions of this product

Catalog No. AAA1012003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 66176-39-4
    Molecular Formula C7H6BrClO2S
    Formula Weight 269.55
    MDL Number MFCD00156129
    UN Number UN3261
    Beilstein 2364328
    Synonym alpha-Bromo-p-toluenesulfonyl chloride
    Chemical Name or Material 4- (Bromomethyl)benzenesulfonyl chloride

    Specifications

    Formula Weight 269.55
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 66176-39-4
    Synonym alpha-Bromo-p-toluenesulfonyl chloride
    UN Number UN3261
    Beilstein 2364328
    Quantity 1g
    Health Hazard 1 Danger
    Molecular Formula C7H6BrClO2S
    MDL Number MFCD00156129
    Melting Point 72° to 75°C
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4- (Bromomethyl)benzenesulfonyl chloride
    Assay 95%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit