Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromomethyl)biphenyl, 96%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
2567-29-5
C13H11Br
247.13
HZQLUIZFUXNFHK-UHFFFAOYSA-N
4-bromomethylbiphenyl,4-bromomethyl biphenyl,4-bromomethyl-1,1'-biphenyl,4-phenylbenzyl bromide,1-bromomethyl-4-phenylbenzene,bromoethylbiphenyl,4-biphenylmethylbromide,4-bromomethyl-biphenyl
257716
1-(bromomethyl)-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)CBr
2567-29-5
C13H11Br
247.13
HZQLUIZFUXNFHK-UHFFFAOYSA-N
4-bromomethylbiphenyl,4-bromomethyl biphenyl,4-bromomethyl-1,1'-biphenyl,4-phenylbenzyl bromide,1-bromomethyl-4-phenylbenzene,bromoethylbiphenyl,4-biphenylmethylbromide,4-bromomethyl-biphenyl
257716
1-(bromomethyl)-4-phenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)CBr
Melting Point 83.0°C to 86.0°C
Quantity 100g
Flash Point >110°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula C6H5C6H4CH2Br
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 247.13
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 4-(Bromomethyl)biphenyl
Show More Show Less

GHS Signal Word: Danger