Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethylbenzenesulfonyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
66176-39-4
C7H6BrClO2S
269.55
MFCD00156129
QXTQWYZHHMQSQH-UHFFFAOYSA-N
4-bromomethyl benzenesulfonyl chloride,4-bromomethyl benzene-1-sulfonyl chloride,4-bromomethyl benzenesulfonylchloride,alpha-bromo-p-toluenesulphonyl chloride,alpha-bromo-p-toluenesulfonyl chloride,4-bromomethyl benzenesulphonyl chloride,4-bromomethyl benzene-1-sulfonylchloride,buttpark 94\04-08,4-bromomethyl phenyl chlorosulfone,4-bromomethylene benzenesulfonyl chloride
2734409
4-(bromomethyl)benzenesulfonyl chloride
C1=CC(=CC=C1CBr)S(=O)(=O)Cl
66176-39-4
C7H6BrClO2S
269.55
MFCD00156129
QXTQWYZHHMQSQH-UHFFFAOYSA-N
4-bromomethyl benzenesulfonyl chloride,4-bromomethyl benzene-1-sulfonyl chloride,4-bromomethyl benzenesulfonylchloride,alpha-bromo-p-toluenesulphonyl chloride,alpha-bromo-p-toluenesulfonyl chloride,4-bromomethyl benzenesulphonyl chloride,4-bromomethyl benzene-1-sulfonylchloride,buttpark 94\04-08,4-bromomethyl phenyl chlorosulfone,4-bromomethylene benzenesulfonyl chloride
2734409
4-(bromomethyl)benzenesulfonyl chloride
C1=CC(=CC=C1CBr)S(=O)(=O)Cl
Color White to Yellow
Infrared Spectrum Authentic
Quantity 25g
Melting Point 69.0°C to 76.0°C
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula BrCH2C6H4SO2Cl
Formula Weight 269.55
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 4-Bromomethylbenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger