Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenethyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
4654-39-1
C8H9BrO
201.06
MFCD00002897
PMOSJSPFNDUAFY-UHFFFAOYSA-N
2-4-bromophenyl ethanol,4-bromophenethyl alcohol,p-bromophenethyl alcohol,benzeneethanol, 4-bromo,2-4-bromophenyl ethan-1-ol,2-4-bromo-phenyl-ethanol,2-4-bromophenyl ethyl alcohol,2-p-bromophenyl ethanol,phenethyl alcohol, p-bromo,4-bromobenzene ethanol
72851
2-(4-bromophenyl)ethanol
C1=CC(=CC=C1CCO)Br
4654-39-1
C8H9BrO
201.06
MFCD00002897
PMOSJSPFNDUAFY-UHFFFAOYSA-N
2-4-bromophenyl ethanol,4-bromophenethyl alcohol,p-bromophenethyl alcohol,benzeneethanol, 4-bromo,2-4-bromophenyl ethan-1-ol,2-4-bromo-phenyl-ethanol,2-4-bromophenyl ethyl alcohol,2-p-bromophenyl ethanol,phenethyl alcohol, p-bromo,4-bromobenzene ethanol
72851
2-(4-bromophenyl)ethanol
C1=CC(=CC=C1CCO)Br
Specific Gravity 1.43
Boiling Point 96°C to 99°C (0.5mmHg)
Flash Point >110°C
Density 1.4300g/mL
Refractive Index 1.5720 to 1.574
Quantity 10g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula BrC6H4CH2CH2OH
Beilstein 06, III, 1713
Formula Weight 201.06
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Bromophenethyl alcohol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-093-7