Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
73918-56-6
C8H10BrN
200.08
MFCD00008189
ZSZCXAOQVBEPME-UHFFFAOYSA-N
4-bromophenethylamine,2-4-bromophenyl ethylamine,2-4-bromophenyl ethanamine,p-bromophenethylamine,2-4-bromo-phenyl-ethylamine,benzeneethanamine, 4-bromo,2-4-bromophenyl ethan-1-amine,4-bromophenylethylamine,4-bromobenzeneethanamine,1-amino-2-4-bromophenyl ethane
533915
2-(4-bromophenyl)ethanamine
C1=CC(=CC=C1CCN)Br
73918-56-6
C8H10BrN
200.08
MFCD00008189
ZSZCXAOQVBEPME-UHFFFAOYSA-N
4-bromophenethylamine,2-4-bromophenyl ethylamine,2-4-bromophenyl ethanamine,p-bromophenethylamine,2-4-bromo-phenyl-ethylamine,benzeneethanamine, 4-bromo,2-4-bromophenyl ethan-1-amine,4-bromophenylethylamine,4-bromobenzeneethanamine,1-amino-2-4-bromophenyl ethane
533915
2-(4-bromophenyl)ethanamine
C1=CC(=CC=C1CCN)Br
Infrared Spectrum Authentic
Quantity 25mL
Flash Point >110°C
Boiling Point 63°C to 72°C (0.2mmHg)
Specific Gravity 1.29
Color Colorless to Yellow
Packaging Glass bottle
Density 1.2900g/mL
Refractive Index 1.5730 to 1.575
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H4CH2CH2NH2
Formula Weight 200.08
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Bromophenethylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 277-636-2