Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenethyl bromide, 96%, Acros Organics

Click to view available options
Quantity:
1g
10g
1746-28-7
C8H8Br2
263.96
APTDRDYSJZQPPI-UHFFFAOYSA-N
1-bromo-4-2-bromoethyl benzene,4-bromophenethyl bromide,4-bromophenethylbromide,benzene,1-bromo-4-2-bromoethyl,4-bromo-1-2-bromoethyl benzene,pubchem16698,4-bromobenzylmethyl bromide,acmc-209e8m,1-bromo-4-2-bromoethyl ;benzene
11010825
1-bromo-4-(2-bromoethyl)benzene
C1=CC(=CC=C1CCBr)Br
1746-28-7
C8H8Br2
263.96
APTDRDYSJZQPPI-UHFFFAOYSA-N
1-bromo-4-2-bromoethyl benzene,4-bromophenethyl bromide,4-bromophenethylbromide,benzene,1-bromo-4-2-bromoethyl,4-bromo-1-2-bromoethyl benzene,pubchem16698,4-bromobenzylmethyl bromide,acmc-209e8m,1-bromo-4-2-bromoethyl ;benzene
11010825
1-bromo-4-(2-bromoethyl)benzene
C1=CC(=CC=C1CCBr)Br
Boiling Point 97°C to 98.5°C (1.0mmHg)
Flash Point >110°C
Specific Gravity 1.73
Density 1.7300g/mL
Color Colorless to Yellow
Quantity 10g
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 263.96
Physical Form Liquid or Low Melting Solid
Percent Purity 96%
Chemical Name or Material 4-Bromophenethyl bromide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning