Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenetole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
588-96-5
C8H9BrO
201.06
MFCD00000098
WVUYYXUATWMVIT-UHFFFAOYSA-N
4-bromophenetole,p-bromophenetole,benzene, 1-bromo-4-ethoxy,p-bromoethoxybenzene,p-ethoxybromobenzene,p-ethoxyphenyl bromide,p-bromophenol ethyl ether,4-bromophenyl ethyl ether,1-bromo-4-ethoxy-benzene,4-bromoethoxybenzene
68523
1-bromo-4-ethoxybenzene
CCOC1=CC=C(C=C1)Br
588-96-5
C8H9BrO
201.06
MFCD00000098
WVUYYXUATWMVIT-UHFFFAOYSA-N
4-bromophenetole,p-bromophenetole,benzene, 1-bromo-4-ethoxy,p-bromoethoxybenzene,p-ethoxybromobenzene,p-ethoxyphenyl bromide,p-bromophenol ethyl ether,4-bromophenyl ethyl ether,1-bromo-4-ethoxy-benzene,4-bromoethoxybenzene
68523
1-bromo-4-ethoxybenzene
CCOC1=CC=C(C=C1)Br
Flash Point 103°C
Specific Gravity 1.4
Color Colorless to Yellow
Refractive Index 1.5500 to 1.5520
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 233.0°C
Melting Point 4.0°C
Density 1.4000g/mL
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4OC2H5
Beilstein 06, 199
Merck Index 10, 1400
Solubility Information Solubility in water: .. Other solubilities: insoluble in most solvents
Formula Weight 201.06
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Bromophenetole, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-629-7

TSCA : TSCA