Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenyl methyl sulfone, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3466-32-8
C7H7BrO2S
235.1
MFCD00025065
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
4-bromophenyl methyl sulfone,1-bromo-4-methylsulfonyl benzene,1-bromo-4-methanesulfonylbenzene,p-bromophenyl methyl sulfone,methyl 4-bromophenyl sulfone,4-bromophenyl methyl sulphone,benzene, 1-bromo-4-methylsulfonyl,4-bromophenylmethylsulfone,p-methyl benzenesulfobromide,sulfone, p-bromophenyl methyl
77014
1-bromo-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)Br
3466-32-8
C7H7BrO2S
235.1
MFCD00025065
FJLFSYRGFJDJMQ-UHFFFAOYSA-N
4-bromophenyl methyl sulfone,1-bromo-4-methylsulfonyl benzene,1-bromo-4-methanesulfonylbenzene,p-bromophenyl methyl sulfone,methyl 4-bromophenyl sulfone,4-bromophenyl methyl sulphone,benzene, 1-bromo-4-methylsulfonyl,4-bromophenylmethylsulfone,p-methyl benzenesulfobromide,sulfone, p-bromophenyl methyl
77014
1-bromo-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)Br
Quantity 1g
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 102°C to 105°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 235.1
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Bromophenyl methyl sulfone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.