Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromophenylacetic acid, 99%

Click to view available options
Quantity:
5g
25g
100g
1878-68-8
C8H7BrO2
215.046
MFCD00004342
QOWSWEBLNVACCL-UHFFFAOYSA-N
4-bromophenylacetic acid,2-4-bromophenyl acetic acid,4-bromophenyl acetic acid,p-bromophenylacetic acid,4-bromobenzeneacetic acid,benzeneacetic acid, 4-bromo,p-bromophenyl acetic acid,acetic acid, p-bromophenyl,p-bromphenylessigsaure german
74654
CHEBI:1790
2-(4-bromophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)Br
1878-68-8
C8H7BrO2
215.046
MFCD00004342
QOWSWEBLNVACCL-UHFFFAOYSA-N
4-bromophenylacetic acid,2-4-bromophenyl acetic acid,4-bromophenyl acetic acid,p-bromophenylacetic acid,4-bromobenzeneacetic acid,benzeneacetic acid, 4-bromo,p-bromophenyl acetic acid,acetic acid, p-bromophenyl,p-bromphenylessigsaure german
74654
CHEBI:1790
2-(4-bromophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)Br
Quantity 100g
Melting Point 115°C to 119°C
Beilstein 972146
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenylacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 217-523-7

RTECSNumber : AF6325000

TSCA : Yes

Recommended Storage : Ambient temperatures