Alfa Aesar™ 4-Bromophenylacetonitrile, 97%

Manufacturer: Alfa Aesar™ A1088736

Quantity 500g

 View more versions of this product

Catalog No. AAA1088736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16532-79-9
    Molecular Formula C8H6BrN
    Formula Weight 196.05
    MDL Number MFCD00001916
    UN Number UN3449
    Beilstein 1861069
    Synonym 4-Bromobenzyl cyanide
    Chemical Name or Material 4-Bromophenylacetonitrile

    Specifications

    Formula Weight 196.05
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P361-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    MDL Number MFCD00001916
    CAS 16532-79-9
    Melting Point 45° to 49°C
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BROMOBENZYL CYANIDES, SOLID
    Molecular Formula C8H6BrN
    UN Number UN3449
    Synonym 4-Bromobenzyl cyanide
    Flash Point 127°C (261°F)
    Odor Pungent
    Beilstein 1861069
    Boiling Point 286°C
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number AL8090000
    Chemical Name or Material 4-Bromophenylacetonitrile
    Assay 97%

    GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P361-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit