Learn More
Medchemexpress LLC 4-Bromophenylacetylene | 766-96-1 | 181.03 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00754210MM/1ML
Encompass_Preferred
1-Bromo-4-ethynylbenzene is a biochemical reagent suitable for life science-related research, functioning as both a biological material and an organic compound. It serves as a click chemistry reagent, possessing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Biochemical reagent for life science research.
  • Usable as a biological material or organic compound.
  • Acts as a click chemistry reagent.
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
  • Serves as a starting material for creating second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.
Catalog No. 50-004-45812