Learn More
Medchemexpress LLC 4-Bromophenylacetylene | 766-96-1 | 181.03 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0075425G
Encompass_Preferred
1-Bromo-4-ethynylbenzene is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. This compound is also used as a starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes. It is intended for research use only and not sold to patients.
  • Biochemical reagent for life science research
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Starting material for second-order nonlinear optical materials
  • Starting material for heterocyclotriynes and unsymmetrical 1,4-diarylbutadiynes
Catalog No. 50-004-46590