Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromophenylhydrazine hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
622-88-8
C6H8BrClN2
223.498
MFCD00012941
RGGOWBBBHWTTRE-UHFFFAOYSA-N
4-bromophenylhydrazine hydrochloride,4-bromophenyl hydrazine hydrochloride,4-bromophenylhydrazine hcl,4-bromo phenyl hydrazine hydrochloride,p-bromophenylhydrazine monohydrochloride,hydrazine, 4-bromophenyl-, monohydrochloride,1-4-bromophenyl hydrazine hydrochloride,4-bromophenylhydrazinehydrochloride,p-bromophenylhydrazine hydrochloride
12157
(4-bromophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)Br.Cl
622-88-8
C6H8BrClN2
223.498
MFCD00012941
RGGOWBBBHWTTRE-UHFFFAOYSA-N
4-bromophenylhydrazine hydrochloride,4-bromophenyl hydrazine hydrochloride,4-bromophenylhydrazine hcl,4-bromo phenyl hydrazine hydrochloride,p-bromophenylhydrazine monohydrochloride,hydrazine, 4-bromophenyl-, monohydrochloride,1-4-bromophenyl hydrazine hydrochloride,4-bromophenylhydrazinehydrochloride,p-bromophenylhydrazine hydrochloride
12157
(4-bromophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)Br.Cl
Applications
4-Bromophenylhydrazine hydrochloride is used as a reagent in the preparation of acylsulfonamides, acylsulfamides and 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Odor Odorless
Melting Point ~215°C (decomposition)
Quantity 25g
Beilstein 3565838
Solubility Information Soluble in water.
Formula Weight 223.51
Percent Purity 97%
Chemical Name or Material 4-Bromophenylhydrazine hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 255-590-4

RTECSNumber : MV0800000

TSCA : Yes

Recommended Storage : Ambient temperatures