Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromopiperidine hydrobromide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
54288-70-9
C5H10BrN·HBr
244.96
MFCD00191858
LVTIZXGNLIKUQZ-UHFFFAOYSA-N
4-bromopiperidine hydrobromide,4-bromopiperidinehbr,4-bromopiperidine hbr,piperidine, 4-bromo-, hydrobromide,4-bromopiperidine hydrobromide 1:1,4-bromopiperidine, bromide,pubchem9060,acmc-209lf5,4-bromo-piperidine hbr salt,4-bromopiperidinehydrobromide
2734676
4-bromopiperidine;hydrobromide
C1CNCCC1Br.Br
54288-70-9
C5H10BrN·HBr
244.96
MFCD00191858
LVTIZXGNLIKUQZ-UHFFFAOYSA-N
4-bromopiperidine hydrobromide,4-bromopiperidinehbr,4-bromopiperidine hbr,piperidine, 4-bromo-, hydrobromide,4-bromopiperidine hydrobromide 1:1,4-bromopiperidine, bromide,pubchem9060,acmc-209lf5,4-bromo-piperidine hbr salt,4-bromopiperidinehydrobromide
2734676
4-bromopiperidine;hydrobromide
C1CNCCC1Br.Br
Color White
Infrared Spectrum Authentic
Melting Point 199.0°C to 202.0°C
Packaging Glass bottle
Quantity 10g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5% (Argentometry)
Beilstein 20, IV, 1381
Formula Weight 244.96
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Bromopiperidine hydrobromide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.