Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromopyridine-3-carboxylic acid, 96%

Click to view available options
Quantity:
1g
5g
15366-62-8
C6H4BrNO2
202.007
MFCD04114236
CZLOEKRJQIAKFI-UHFFFAOYSA-N
4-bromonicotinic acid,4-bromo-nicotinic acid,3-acid-4-bromopyridine,3-pyridinecarboxylic acid, 4-bromo,4-bromo-nicotinicacid,4-bromo-3-pyridinecarboxylic acid,4-bromonicotinicacid,3-acid-4-bromopyridine,pubchem18140,acmc-209d9s
2762915
4-bromopyridine-3-carboxylic acid
C1=CN=CC(=C1Br)C(=O)O
15366-62-8
C6H4BrNO2
202.007
MFCD04114236
CZLOEKRJQIAKFI-UHFFFAOYSA-N
4-bromonicotinic acid,4-bromo-nicotinic acid,3-acid-4-bromopyridine,3-pyridinecarboxylic acid, 4-bromo,4-bromo-nicotinicacid,4-bromo-3-pyridinecarboxylic acid,4-bromonicotinicacid,3-acid-4-bromopyridine,pubchem18140,acmc-209d9s
2762915
4-bromopyridine-3-carboxylic acid
C1=CN=CC(=C1Br)C(=O)O
Melting Point 118°C to 134°C
Quantity 1g
Color Brown
Formula Weight 202
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4-Bromopyridine-3-carboxylic acid