Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-4-Bromostyrene oxide, 98%

Manufacturer:  Alfa Aesar™ H2681806

Quantity 5g

 View more versions of this product

Catalog No. AAH2681806

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
(+/-)-4-Bromostyrene oxide is used for the synthesis of pharmaceuticals, disinfectants, and products for the perfume industry.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

32017-76-8
199.04
UN1325
(+/-)-4-Bromostyrene oxide
C8H7BrO
MFCD01656787
2-(4-Bromophenyl)oxirane

Specifications

32017-76-8
199.04
UN1325
2-(4-Bromophenyl)oxirane
5g
(+/-)-4-Bromostyrene oxide
C8H7BrO
MFCD01656787
26° to 29°C
91° to 93°C (2mmHg)
38°C (100°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H341-H315-H319-H335
Flammable solid.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CY9025000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit