Alfa Aesar™ (+/-)-4-Bromostyrene oxide, 98%

Manufacturer: Alfa Aesar™ H2681814

Quantity 25g

 View more versions of this product

Catalog No. AAH2681814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32017-76-8
    Molecular Formula C8H7BrO
    Formula Weight 199.04
    MDL Number MFCD01656787
    UN Number UN1325
    Synonym 2-(4-Bromophenyl)oxirane
    Chemical Name or Material (+/-)-4-Bromostyrene oxide

    Specifications

    Formula Weight 199.04
    Molecular Formula C8H7BrO
    MDL Number MFCD01656787
    UN Number UN1325
    Synonym 2-(4-Bromophenyl)oxirane
    Boiling Point 91° to 93°C (2mmHg)
    Quantity 25g
    Flash Point 38°C (100°F)
    Health Hazard 1 Warning
    RTECS Number CY9025000
    CAS 32017-76-8
    Melting Point 26° to 29°C
    Health Hazard 2 GHS H Statement
    H228-H341-H315-H319-H335
    Flammable solid.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material (+/-)-4-Bromostyrene oxide
    Assay 98%

    Description

    Applications
    (+/-)-4-Bromostyrene oxide is used for the synthesis of pharmaceuticals, disinfectants, and products for the perfume industry.

    Solubility
    Insoluble in water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H228-H341-H315-H319-H335
    Flammable solid.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit