Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromostyrene, 96%, stabilized, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
100mL
Packaging:
Glass bottle
2039-82-9
C8H7Br
183.05
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene,1-bromo-4-vinylbenzene,p-bromostyrene,benzene, 1-bromo-4-ethenyl,styrene, p-bromo,1-4-bromophenyl ethylene,unii-tvk4s80xlu,p-bromo styrene,1-bromo-4-vinyl-benzene,tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
C=CC1=CC=C(C=C1)Br
2039-82-9
C8H7Br
183.05
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene,1-bromo-4-vinylbenzene,p-bromostyrene,benzene, 1-bromo-4-ethenyl,styrene, p-bromo,1-4-bromophenyl ethylene,unii-tvk4s80xlu,p-bromo styrene,1-bromo-4-vinyl-benzene,tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
C=CC1=CC=C(C=C1)Br
Melting Point 4.5°C
Stabilizer 100 to 500ppm 4-tert.-butylcatechol
Infrared Spectrum Authentic
Quantity 25mL
Specific Gravity 1.4
Color Colorless to Yellow
Flash Point 75°C
Density 1.4000g/mL
Refractive Index 1.5930 to 1.5950
Boiling Point 88.0°C to 89.0°C (16.0mmHg)
Packaging Glass bottle
Assay Percent Range 95% min. (GC)
Linear Formula H2C=CHC6H4Br
Beilstein 05,II,367
Formula Weight 183.05
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 4-Bromostyrene
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 218-022-6