Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H5595614

Quantity 25g

 View more versions of this product

Catalog No. AAH5595614


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

26197-93-3
216.1
MFCD04114360
4-Bromothiobenzamide

Specifications

26197-93-3
216.1
White to pale brown
25g
97%
Solid
MFCD04114360
139° to 143°C
4-Bromothiobenzamide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit