Alfa Aesar™ 4'-Bromovalerophenone, 98%

Manufacturer: Alfa Aesar™ B2497906

Quantity 5g

 View more versions of this product

Catalog No. AAB2497906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7295-44-5
    Molecular Formula C11H13BrO
    Formula Weight 241.14
    MDL Number MFCD00000107
    Synonym 4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
    Chemical Name or Material 4'-Bromovalerophenone

    Specifications

    Formula Weight 241.14
    Molecular Formula C11H13BrO
    MDL Number MFCD00000107
    Synonym 4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
    Melting Point 34° to 36°C
    Quantity 5g
    Flash Point >110°C (230°F)
    CAS 7295-44-5
    Boiling Point 168° to 169°C (20mmHg)
    Chemical Name or Material 4'-Bromovalerophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit