Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Bromovalerophenone, 98%

Manufacturer:  Alfa Aesar™ B2497922

Quantity 100g

 View more versions of this product

Catalog No. AAB2497922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7295-44-5
241.14
4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
C11H13BrO
MFCD00000107
4'-Bromovalerophenone

Specifications

7295-44-5
241.14
4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
168° to 169°C (20mmHg)
>110°C (230°F)
98%
C11H13BrO
MFCD00000107
34° to 36°C
100g
4'-Bromovalerophenone
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit