Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Bromovalerophenone, 98%

Click to view available options
Quantity:
5g
25g
100g
7295-44-5
C11H13BrO
241.128
MFCD00000107
STYJKBMRWQQJIS-UHFFFAOYSA-N
4'-bromovalerophenone,1-4-bromophenyl pentan-1-one,p-bromophenyl butyl ketone,p-bromovalerophenone,1-pentanone, 1-4-bromophenyl,4-bromovalerophenone,1-bromo-4-pentanoylbenzene,1-4-bromophenyl-1-pentanone,acmc-20ammf,4;-bromovalerophenone
81717
1-(4-bromophenyl)pentan-1-one
CCCCC(=O)C1=CC=C(C=C1)Br
7295-44-5
C11H13BrO
241.128
MFCD00000107
STYJKBMRWQQJIS-UHFFFAOYSA-N
4'-bromovalerophenone,1-4-bromophenyl pentan-1-one,p-bromophenyl butyl ketone,p-bromovalerophenone,1-pentanone, 1-4-bromophenyl,4-bromovalerophenone,1-bromo-4-pentanoylbenzene,1-4-bromophenyl-1-pentanone,acmc-20ammf,4;-bromovalerophenone
81717
1-(4-bromophenyl)pentan-1-one
CCCCC(=O)C1=CC=C(C=C1)Br
Flash Point >110°C (230°F)
Quantity 25g
Melting Point 34°C to 36°C
Boiling Point 168°C to 169°C (20mmHg)
Formula Weight 241.14
Percent Purity 98%
Chemical Name or Material 4'-Bromovalerophenone