Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromoveratrole, 97%

Manufacturer:  Alfa Aesar™ A1113322

Quantity 100g

 View more versions of this product

Catalog No. AAA1113322


  Check Availability
Add to cart

Specifications

Description

Applications
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

Solubility
Soluble in benzene, ether, toluene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

2859-78-1
217.06
2046884
4-Bromoveratrole
C8H9BrO2
MFCD00008381
4-Bromo-1,2-dimethoxybenzene; 3,4-Dimethoxybromobenzene

Specifications

2859-78-1
217.06
2046884
255° to 256°C
109°C (228°F)
97%
C8H9BrO2
MFCD00008381
4-Bromo-1,2-dimethoxybenzene; 3,4-Dimethoxybromobenzene
100g
4-Bromoveratrole

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit