Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid,benzoic acid, 4-butyl,4-butyl-benzoic acid,p-butylbenzoic acid,p-n-butylbenzoic acid,p-butyl benzoic acid,4-butylbenzoicacid,p-n-butyl benzoic acid,acmc-209fcp,4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid,benzoic acid, 4-butyl,4-butyl-benzoic acid,p-butylbenzoic acid,p-n-butylbenzoic acid,p-butyl benzoic acid,4-butylbenzoicacid,p-n-butyl benzoic acid,acmc-209fcp,4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color White
Melting Point 100°C to 103°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)3C6H4COOH
Beilstein 09, III, 2520
Formula Weight 178.23
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Butylbenzoic acid, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-940-9

TSCA : TSCA