Alfa Aesar™ 4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester, 96%

Manufacturer: Alfa Aesar™ H52316MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52316MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C13H16BF2NO3
    Formula Weight 283.08
    MDL Number MFCD17215212
    Synonym 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
    Chemical Name or Material 4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester

    Specifications

    Formula Weight 283.08
    Molecular Formula C13H16BF2NO3
    MDL Number MFCD17215212
    Synonym 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
    Quantity 250mg
    Chemical Name or Material 4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit