Alfa Aesar™ 4-Carboxy-2-nitrobenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H5275703

Quantity 1g

 View more versions of this product

Catalog No. AAH5275703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85107-54-6
    Molecular Formula C7H6BNO6
    Formula Weight 210.94
    MDL Number MFCD00448780
    Synonym 4-Carboxy-2-nitrophenylboronic acid; 4-Borono-3-nitrobenzoic acid
    Chemical Name or Material 4-Carboxy-2-nitrobenzeneboronic acid

    Specifications

    Formula Weight 210.94
    Molecular Formula C7H6BNO6
    MDL Number MFCD00448780
    Synonym 4-Carboxy-2-nitrophenylboronic acid; 4-Borono-3-nitrobenzoic acid
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    CAS 85107-54-6
    Health Hazard 1 Warning
    Melting Point 265° to 268°C
    Chemical Name or Material 4-Carboxy-2-nitrobenzeneboronic acid
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit