Alfa Aesar™ 4-Carboxybenzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H2682503

Quantity 1g

 View more versions of this product

Catalog No. AAH2682503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 180516-87-4
    Molecular Formula C13H17BO4
    Formula Weight 248.08
    MDL Number MFCD01863710
    Synonym 4-Carboxylphenylboronic acid pinacol ester; 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
    Chemical Name or Material 4-Carboxybenzeneboronic acid pinacol ester

    Specifications

    Formula Weight 248.08
    Molecular Formula C13H17BO4
    MDL Number MFCD01863710
    CAS 180516-87-4
    Synonym 4-Carboxylphenylboronic acid pinacol ester; 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
    Melting Point 227° to 231°C
    Quantity 1g
    Chemical Name or Material 4-Carboxybenzeneboronic acid pinacol ester
    Assay 97%

    Description

    Applications
    4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit