Learn More
4-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Description
4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 227°C to 231°C |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 248.08 |
Percent Purity | 97% |
Chemical Name or Material | 4-Carboxybenzeneboronic acid pinacol ester |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.