Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(4-Carboxybutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.
Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.
Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
Specifications
Melting Point | 205°C to 209°C |
Flash Point | 195°C (383°F) |
Linear Formula | HO2C(CH2)4P(C6H5)3Br |
Quantity | 25 g |
Beilstein | 3586477 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in ethanol,methanol and soluble water. Insoluble in toluene and hexane. |
Formula Weight | 443.33 |
Percent Purity | 98% |
Chemical Name or Material | (4-Carboxybutyl)triphenylphosphonium bromide |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 241-782-5 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction