Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Carboxymethyl)phenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
96% min. (HPLC)
C8H9BO4
4-(Carboxymethyl)phenylboronic acid
96% min. (HPLC)
C8H9BO4
4-(Carboxymethyl)phenylboronic acid
Packaging Glass Bottle
Quantity 1g
CAS 90111-58-3
Synonym 2-4-boronophenyl acetic acid,4-carboxymethylphenylboronic acid,4-carboxymethyl phenylboronic acid,benzeneacetic acid, 4-borono,4-boronobenzeneacetic acid,4-boronophenylacetic acid,4-carboxymethyl-phenylboronic acid,4-dihydroxyboryl phenyl acetic acid,4-acetic acid-benzeneboronic acid,4-carboxymethyl benzeneboronic acid
InChI Key NFGJQVPDPIGBJE-UHFFFAOYSA-N
Molecular Weight (g/mol) 179.97
PubChem CID 21878961
Formula Weight 179.97
Percent Purity 97%

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning