Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1,8-naphthalic anhydride, Tech., ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
4053-08-1
C12H5ClO3
232.62
MFCD00006928
UJEUBSWHCGDJQU-UHFFFAOYSA-N
4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione
77672
C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl
4053-08-1
C12H5ClO3
232.62
MFCD00006928
UJEUBSWHCGDJQU-UHFFFAOYSA-N
4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione
77672
C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl
Color Beige to Brown
Infrared Spectrum Authentic
Loss on Drying 50% max.
Packaging Plastic bottle
Quantity 250g
Beilstein 17, 522
Formula Weight 232.62
Physical Form Powder
Grade Technical
Chemical Name or Material 4-Chloro-1, 8-naphthalic anhydride, tech.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-760-7

RTECSNumber : QL6127295

TSCA : TSCA