Alfa Aesar™ 4-Chloro-1H-indazole, 97%

Manufacturer: Alfa Aesar™ H3741603

Quantity 1g

 View more versions of this product

Catalog No. AAH3741603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13096-96-3
    Formula Weight 152.58
    MDL Number MFCD00211065
    UN Number UN2811
    Chemical Name or Material 4-Chloro-1H-indazole

    Specifications

    Formula Weight 152.58
    Melting Point 155° to 160°C
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    MDL Number MFCD00211065
    CAS 13096-96-3
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 4-Chloro-1H-indazole
    Assay 97%

    GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit