Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2,5-difluorobenzaldehyde, 98%

Click to view available options
Quantity:
250mg
1g
5g
879093-02-4
C7H3ClF2O
176.547
MFCD07782051
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluoro-benzaldehyde,benzaldehyde, 4-chloro-2,5-difluoro,2,5-difluoro-4-chlorobenzaldehyde,pubchem1430,benzaldehyde,4-chloro-2,5-difluoro
18991142
4-chloro-2,5-difluorobenzaldehyde
C1=C(C(=CC(=C1F)Cl)F)C=O
879093-02-4
C7H3ClF2O
176.547
MFCD07782051
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluoro-benzaldehyde,benzaldehyde, 4-chloro-2,5-difluoro,2,5-difluoro-4-chlorobenzaldehyde,pubchem1430,benzaldehyde,4-chloro-2,5-difluoro
18991142
4-chloro-2,5-difluorobenzaldehyde
C1=C(C(=CC(=C1F)Cl)F)C=O
Quantity 250mg
Color White
Beilstein 14516595
Formula Weight 176.55
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,5-difluorobenzaldehyde

TSCA : No

Recommended Storage : Ambient temperatures