Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2,5-dimethylbenzenesulfonamide, 97%

Click to view available options
Quantity:
1g
5g
219689-73-3
C8H10ClNO2S
219.683
MFCD00275263
YSDCIOQWHXYICL-UHFFFAOYSA-N
4-chloro-2,5-dimethylbenzene-1-sulfonamide,acmc-1chyl,5-chloro-2-sulphamoyl-p-xylene,4-chloro-2,5-dimethylbenzenesulphonamide,benzenesulfonamide,4-chloro-2,5-dimethyl,4-chloranyl-2,5-dimethyl-benzenesulfonamide
2735730
4-chloro-2,5-dimethylbenzenesulfonamide
CC1=CC(=C(C=C1Cl)C)S(=O)(=O)N
219689-73-3
C8H10ClNO2S
219.683
MFCD00275263
YSDCIOQWHXYICL-UHFFFAOYSA-N
4-chloro-2,5-dimethylbenzene-1-sulfonamide,acmc-1chyl,5-chloro-2-sulphamoyl-p-xylene,4-chloro-2,5-dimethylbenzenesulphonamide,benzenesulfonamide,4-chloro-2,5-dimethyl,4-chloranyl-2,5-dimethyl-benzenesulfonamide
2735730
4-chloro-2,5-dimethylbenzenesulfonamide
CC1=CC(=C(C=C1Cl)C)S(=O)(=O)N
Melting Point 190°C to 192°C
Quantity 5g
Formula Weight 219.69
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,5-dimethylbenzenesulfonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning