Learn More
Alfa Aesar™ 4-Chloro-2-fluoro-3-methoxybenzylamine, 97%
Manufacturer:  Alfa Aesar™ H33936MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH33936MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
C8H9ClFNO
MFCD19687209
4-Chloro-2-fluoro-3-methoxybenzylamine
189.61
UN3259
Specifications
250mg
189.61
UN3259
97%
C8H9ClFNO
MFCD19687209
4-Chloro-2-fluoro-3-methoxybenzylamine
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.