Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-fluorocinnamic acid, 98%

Click to view available options
Quantity:
5g
25g
202982-65-8
C9H6ClFO2
200.593
MFCD00143287
FVLPOWWRHAOKMT-DUXPYHPUSA-N
4-chloro-2-fluorocinnamic acid,3-4-chloro-2-fluorophenyl acrylic acid,2e-3-4-chloro-2-fluorophenyl prop-2-enoic acid,4-chloro-2-fluorocinnamicacid,3-4-chloro-2-fluorophenyl prop-2-enoic acid,pubchem3431,e-3-4-chloro-2-fluorophenyl acrylic acid,rarechem bk hw 0062,timtec-bb sbb003542,trans-4-chloro-2-fluorocinnamic acid
5941651
(E)-3-(4-chloro-2-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1Cl)F)C=CC(=O)O
202982-65-8
C9H6ClFO2
200.593
MFCD00143287
FVLPOWWRHAOKMT-DUXPYHPUSA-N
4-chloro-2-fluorocinnamic acid,3-4-chloro-2-fluorophenyl acrylic acid,2e-3-4-chloro-2-fluorophenyl prop-2-enoic acid,4-chloro-2-fluorocinnamicacid,3-4-chloro-2-fluorophenyl prop-2-enoic acid,pubchem3431,e-3-4-chloro-2-fluorophenyl acrylic acid,rarechem bk hw 0062,timtec-bb sbb003542,trans-4-chloro-2-fluorocinnamic acid
5941651
(E)-3-(4-chloro-2-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1Cl)F)C=CC(=O)O
Melting Point ~215°C
Quantity 25g
Formula Weight 200.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures