Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorophenol, 98+%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol,phenol, 4-chloro-2-fluoro,4-chloro-2-fluoro-phenol,4-chloro-fluorophenol,pubchem2861,acmc-1ct1y,ksc222g4r,4-chloro-2-fluorophenol,attercop-chm at106713
181975
4-chloro-2-fluorophenol
C1=CC(=C(C=C1Cl)F)O
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol,phenol, 4-chloro-2-fluoro,4-chloro-2-fluoro-phenol,4-chloro-fluorophenol,pubchem2861,acmc-1ct1y,ksc222g4r,4-chloro-2-fluorophenol,attercop-chm at106713
181975
4-chloro-2-fluorophenol
C1=CC(=C(C=C1Cl)F)O
Flash Point 81°C
Melting Point 20°C
Boiling Point 103°C to 104°C (50.0mmHg)
Refractive Index 1.5330 to 1.535
Specific Gravity 1.378
Density 1.3780g/mL
Color Colorless to Pink
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula ClC6H3FOH
Solubility Information Solubility in water: insoluble
Formula Weight 146.55
Physical Form Liquid or Low Melting Solid
Percent Purity 98+%
Chemical Name or Material 4-Chloro-2-fluorophenol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning