Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-fluoropyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
34941-92-9
C5H3ClFN
131.534
MFCD04112503
GNJKJKBURMCLOR-UHFFFAOYSA-N
2-fluoro-4-chloropyridine,4-chloro-2-fluoro-pyridine,pyridine, 4-chloro-2-fluoro,pubchem13535,pyridine,4-chloro-2-fluoro
2762839
4-chloro-2-fluoropyridine
C1=CN=C(C=C1Cl)F
34941-92-9
C5H3ClFN
131.534
MFCD04112503
GNJKJKBURMCLOR-UHFFFAOYSA-N
2-fluoro-4-chloropyridine,4-chloro-2-fluoro-pyridine,pyridine, 4-chloro-2-fluoro,pubchem13535,pyridine,4-chloro-2-fluoro
2762839
4-chloro-2-fluoropyridine
C1=CN=C(C=C1Cl)F
Quantity 250mg
Beilstein 1634727
Formula Weight 131.53
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Chloro-2-fluoropyridine