Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-2'-hydroxyacetophenone, 97%, Acros Organics

Click to view available options
Quantity:
10g
50g
6921-66-0
C8H7ClO2
170.59
QCVSDCHNBNFJDQ-UHFFFAOYSA-N
1-4-chloro-2-hydroxyphenyl ethanone,4'-chloro-2'-hydroxyacetophenone,4-chloro-2-hydroxyacetophenone,1-4-chloro-2-hydroxyphenyl ethan-1-one,2'-hydroxy-4'-chloroacetophenone,1-4-chloro-2-hydroxy-phenyl ethanone,ethanone, 1-4-chloro-2-hydroxyphenyl,4'-chloro-2'-hydroxy-acetophenone,ethanone,1-4-chloro-2-hydroxyphenyl
1051513
1-(4-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)O
6921-66-0
C8H7ClO2
170.59
QCVSDCHNBNFJDQ-UHFFFAOYSA-N
1-4-chloro-2-hydroxyphenyl ethanone,4'-chloro-2'-hydroxyacetophenone,4-chloro-2-hydroxyacetophenone,1-4-chloro-2-hydroxyphenyl ethan-1-one,2'-hydroxy-4'-chloroacetophenone,1-4-chloro-2-hydroxy-phenyl ethanone,ethanone, 1-4-chloro-2-hydroxyphenyl,4'-chloro-2'-hydroxy-acetophenone,ethanone,1-4-chloro-2-hydroxyphenyl
1051513
1-(4-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)O
Melting Point 27°C to 30°C
Quantity 10g
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula CLC6H3(OH)COCH3
Solubility Information Solubility in water: insoluble
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-hydroxyacetophenone

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning